Learn More
Innovative Research Inc Dog Albumin Pur Lyo 200mg
Supplier:  Innovative Research Inc ICNALBLY200MG
Encompass_Preferred
Canine Albumin Purified Lyophilized 200mg - Canine Albumin Purified Lyophilized from Innovative Research has been purified from US origin dog plasma by a propritetary process that combines multiple conventional chromatographic methods. This is a Lyophilized powder buffered in Salt free Lyophilized and a purity of >95% as determined by SDS-PAGE. More Details Species Canine Target Albumin Purification Method Chromatographically purified Purity >95% as determined by SDS-PAGE Analysis Source Dog plasma Storage Conditions 4 C Additional Information Add deionized water or buffer to original or desired volume, aliquot and freeze unused portion. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including blocking, cell signaling, drug studies, protein-protein interactions, protein stabilization, receptor binding, tissue culture, and more.
Catalog No. 50-274-4611