Learn More
Cusabio Technology LLC DOG IL1RN ELISA KIT
Supplier:  Cusabio Technology LLC CSBEL011628DO
Encompass_Preferred
This kit detects IL1RN (Interleukin-1 receptor antagonist protein) in dogs designed for quantitative measurement in serum plasma and tissue homogenates Utilizing a sandwich ELISA method it offers a detection range of 15 6 pg/ml to 1000 pg/ml with a sensitivity of 3 9 pg/ml ensuring precise measurement of low-abundance samples The assay requires a short incubation period of 1-5 hours and uses a modest sample volume of 50-100 l per test Absorbance is measured at 450 nm for result analysis providing reliable and reproducible data Ideal for research applications requiring IL1RN level assessment this kit combines high sensitivity with streamlined protocols to support studies in immunology and inflammatory response monitoring
Catalog No. 50-000-51213