Learn More
Cayman Chemical Dog LIvR X ReceptrAlpha R 96
SDP
Supplier:  Cayman Chemical 4103296 WELL
Encompass_Preferred
INDIGO Bioscience’s assays are all-inclusive cell-based assay systems. In addition to LXRα Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist TO901317, Luciferase Detection Reagent, and a cell culture-ready assay plate.
Catalog No. 50-294-5081