Learn More
Cayman Chemical Dog PRoxIsome ProlIfRatrA 3x32
SDP
Supplier:  Cayman Chemical 283123X32 WELL
Encompass_Preferred
dPPARα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. D00111-32, D00111]
Catalog No. 50-293-9346