Learn More
Immunology Consultants Dog Soluble Interleukin-2 Receptor (sIL-2R) ELISA Kit
SDP
Supplier:  Immunology Consultants E40SLR
Encompass_Preferred
A reliable ELISA Kit for quantifying soluble interleukin-2 receptor (sIL-2R, sIL2R, sTAC, sCD25) in canine/dog serum/plasma samples. It is a highly sensitive two-site enzyme linked immunoassay (ELISA).
Catalog No. 50-240-6525