Learn More
TARGETMOL CHEMICALS INC Dolastatin 10 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TP11801MG
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Dolastatin 10 (DLS 10) is a potent peptide with antimitotic properties, effectively inhibiting tubulin polymerization. Purity 99.55%
Catalog No. 50-287-7312