Learn More
Kingfisher Biotech Inc Dolphin CXCL10 ELISpot - Up to two (2) ELISpot assays
SDP
Supplier:  Kingfisher Biotech Inc SPOT2411P004
Encompass_Preferred
Dolphin CXCL10 ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-289-1931