Learn More
Medchemexpress LLC HY-13238 10mg Medchemexpress, Dolutegravir CAS:1051375-16-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1323810MG
Encompass_Preferred
Medchemexpress, HY-13238 10mg Dolutegravir CAS:1051375-16-6 Dolutegravir (S/GSK1349572) is a highly potent and orally bioavailable HIV integrase strand transfer inhibitor with an IC50 of 2.7 nM for HIV-1 integrase-catalyzed strand transfer. Dolutegravir (S/GSK1349572) inhibits HIV-1 viral replication with an IC50 of 0.51 nM in peripheral blood mononuclear cells. Dolutegravir retains a high potency against the HIV-1 Y143R, N155H, and G140S/Q148H mutants (EC50=3.6-5.8 nM)[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8966