Learn More
Medchemexpress LLC Dolutegravir intermediate-1 | 1335210-23-5 | MFCD28978322 | 99.8% | 315.28 g/mol | C13H17NO8 | 25 G

Supplier: Medchemexpress LLC HY10008325G
Dolutegravir intermediate-1 is a synthetic chemical intermediate used in the synthesis and research of HIV-1 integrase inhibitors. Supplied as a solid with high reported purity and offered in multiple pack sizes, it is intended for laboratory research and chemical development applications.
- High reported purity (99.8%).
- CAS-registered identifier: 1335210-23-5.
- Available as a solid in multiple pack sizes, including 25 g.
- Suitable for use as an intermediate in API synthesis and medicinal chemistry research.
- Safety data sheet and technical documentation available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.