Learn More
TARGETMOL CHEMICALS INC DOMATINOSTAT TOSYLATE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T447710MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Domatinostat tosylate (4SC-202) is a selective inhibitor of class I HDAC for HDAC1/2/3 (IC50 1.20/1.12/0.57 uM). It also displays inhibitory activity against Lysine-specific demethylase 1 (LSD1). purity: 96%
Catalog No. 50-257-9880