Learn More
Cayman Chemical DomIphenbromIde 50g
SDP
Supplier:  Cayman Chemical 3027650 G
Encompass_Preferred
A quaternary ammonium-containing cationic surfactant with antimicrobial activity; active against A. viscosus, A. naeslundii, S. mutans, E. coli, and L. monocytogenes bacteria; inhibits C. neoformans yeast growth and spore germination; inhibits the hERG channel (IC50 = 1.5 μM in a whole-cell patch-clamp assay)
Catalog No. 50-297-8919