Learn More
AdipoGen Domperidone (100 mg)
SDP
Supplier:  AdipoGen CDXD0809M100
Encompass_Preferred
Chemical. CAS: 57808-66-9. Formula: C22H24ClN5O2. MW: 425.91. Domperidone is a peripheral dopamine D2-like receptor antagonist that does not readily cross the blood-brain barrier. It demonstrates a potent affinity for the rat D2 receptor and is also reported to inhibit the rat D3 receptor. The antidopaminergic effects are cross-realted to its gastroprokinetic and antiemetic properties.
Catalog No. 50-205-4403