Learn More
Apexbio Technology LLC Domperidone 57808-66-9 200mg
SDP
Supplier:  Apexbio Technology LLC B1481200
Encompass_Preferred
Domperidone is a small-molecule antagonist targeting the dopamine D2 receptor It is designed to inhibit peripheral dopamine receptors thereby enhancing gastrointestinal motility and accelerating gastric emptying Domperidone exerts its biological activity primarily through peripheral D2 receptor antagonism Based on these pharmacological properties domperidone holds research potential in the investigation and management of gastroparesis and as a comparative compound for mechanistic studies of gastric motility disorders
Catalog No. 50-000-92042