Learn More
Medchemexpress LLC Domperidone (monomaleate) | 83898-65-1 | >99.76% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0411A50MG
Encompass_Preferred
Domperidone monomaleate (R33812) is an orally active and selective dopamine-2 receptor antagonist. It functions as an antiemetic and a prokinetic agent by affecting the chemoreceptor trigger zone and the motor function of the stomach and small intestine. It is rapidly absorbed after oral administration, with few reported side effects.
  • Minimal penetration through the blood-brain barrier
  • Does not cause adverse neurological symptoms
  • Orally active and selective dopamine-2 receptor antagonist
  • Functions as an antiemetic
  • Functions as a prokinetic agent
Catalog No. 50-004-05298