Learn More
Medchemexpress LLC Donepezil-d5 hydrochloride | 1883548-90-0 | C24H25D5ClNO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0034S1MG
Encompass_Preferred
Donepezil-d5 (hydrochloride) is a deuterium labeled version of Donepezil (Hydrochloride), which is a reversible and selective AChE inhibitor with an IC50 of 6.7 nM. It demonstrates high selectivity for AChE over BuChE and has shown neuroprotective effects against Aβ42 neurotoxicity. This product is for research use only and has not been fully validated for medical applications.
  • Stable heavy isotopes used as tracers for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • For research use only
Catalog No. 50-004-32765