Learn More
Medchemexpress LLC Donepezil (E2020 free base) | 120014-06-4 | MFCD00912833 | >98.0% | 379.49 g/mol | C24H29NO3 | 5 MG

Supplier: Medchemexpress LLC HY145665MG
Donepezil (E2020 free base) is a reversible acetylcholinesterase (AChE) inhibitor supplied for research and analytical use. It exhibits potent AChE inhibition (IC50s reported as 8.12 nM for bovine AChE and 11.6 nM for human AChE), with molecular formula C24H29NO3 and molecular weight 379.49 g/mol.
- Potent, specific AChE inhibition useful for pharmacology studies.
- High purity suitable for analytical and research applications.
- Soluble in DMSO (~33.33 mg/mL) and formulatable in common vehicles for dosing.
- Storage stability: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
- Provided in small pack sizes suitable for preparing stock solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.