Learn More
Apexbio Technology LLC Donepezil HCl, 5g. Cas: 120011-70-3 MFCD: MFCD00881312. AChE inhibitor
SDP
Supplier:  Apexbio Technology LLC B16025000
Encompass_Preferred
Donepezil Hydrochloride (Donepezil HCL) is a novel, potent and selective inhibitor of acetylcholinesterase (AChE), an enzyme possibly involved in cognitive dysfunction of patients suffering Alzheimers disease (AD), with the half maximal inhibition concentration IC50 value of 6.7 nM [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5273