Learn More
Selleck Chemical LLC Donepezil HCl S2462-1g
SDP
Supplier:  Selleck Chemical LLC S24621G
Encompass_Preferred
Donepezil HCl is a specific and potent AChE inhibitor for bAChE and hAChE with IC50 of 8 12 nM and 11 6 nM respectively This product is not soluble in PBS solution Please do not dissolve it in PBS for administration
Catalog No. 50-311-1750