Learn More
Medchemexpress LLC Donepezil Hydrochloride | 120011-70-3 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB003450MG
Encompass_Preferred
Donepezil Hydrochloride (E2020) is a reversible, selective AChE inhibitor with an IC50 of 6.7 nM, showing high selectivity for AChE over BuChE. It exhibits a neuroprotective effect on Aβ42 neurotoxicity. In vivo, Donepezil treatment prevents the progression of scopolamine-induced memory impairment in mice, demonstrating its potential in neurodegenerative research.
  • Reversible, selective AChE inhibitor
  • IC50 of 6.7 nM for AChE activity
  • High selectivity for AChE over BuChE
  • Exhibits a neuroprotective effect on Aβ42 neurotoxicity
  • Enhances phosphorylation of Akt and GSK-3β
  • Reduces phosphorylation of tau and glycogen synthase
  • Increases cell viability in cortical neuronal cells
  • Prevents scopolamine-induced memory impairment in mice
Catalog No. 50-004-32082