Learn More
Medchemexpress LLC Donepezil Hydrochloride | 120011-70-3 | 99.9% | C24H30ClNO3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0034100MG
Encompass_Preferred
Donepezil Hydrochloride is a reversible, selective AChE inhibitor with an IC50 of 6.7 nM for AChE activity. It shows high selectivity for AChE over BuChE and exhibits a neuroprotective effect on Aβ42 neurotoxicity.
  • Reversible, selective AChE inhibitor
  • High selectivity for AChE over BuChE
  • Exhibits neuroprotective effect on Aβ42 neurotoxicity
  • For research use only.
Catalog No. 50-004-32503