Learn More
Medchemexpress LLC Donepezil (S)-enantiomer | 142057-80-5 | MFCD00912833 | 379.49 g/mol | C24H29NO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14566A5MG
Encompass_Preferred
(S)-Donepezil is the S-enantiomer of donepezil, a potent and selective acetylcholinesterase (AChE) inhibitor used in research to probe cholinergic signaling and AChE-related pathways. The compound is supplied as a solid and as a 10 mM solution in DMSO and is intended for research use only.
  • Potent acetylcholinesterase inhibition suitable for biochemical and cellular assays.
  • Available as powder and DMSO solution for flexible dosing and preparation.
  • Known chemical formula C24H29NO3 and molecular weight 379.49 g/mol for accurate calculations.
  • Storage recommendations provided to preserve stability (powder and in solvent conditions).
  • Batch-specific purity and documentation available upon request to support experimental reproducibility.
Catalog No. 50-002-21771