Learn More
Medchemexpress LLC Donitriptan | 170912-52-4 | MFCD09832566 | 98.1% | 403.48 g·mol⁻1 | C23H25N5O2 | 10 MG

Supplier: Medchemexpress LLC HY10615710MG
Donitriptan is a research compound characterized as a potent, high-efficacy agonist at serotonin 5-HT1B and 5-HT1D receptors. It was investigated as an antimigraine agent and is intended for laboratory research use only (CAS 170912-52-4).
- Potent agonist at 5-HT1B and 5-HT1D receptors with reported pKi values of 9.4 and 9.3.
- Intended for research use only; not for human or clinical use.
- Molecular formula C23H25N5O2 and molecular weight 403.48 g·mol⁻1.
- Suitable as a reference compound for receptor pharmacology and assay development.
- Purity reported as ~98.1% on distributor listings; manufacturer datasheet does not specify purity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.