Learn More
Medchemexpress LLC Donitriptan | 170912-52-4 | MFCD09832566 | 98.1% | 403.48 g/mol | C23H25N5O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10615750MG
Encompass_Preferred
Donitriptan is a selective serotonin 5-HT1B/1D receptor agonist supplied as a research-grade powder for laboratory studies. It shows high affinity at 5-HT1B (pKi = 9.4) and 5-HT1D (pKi = 9.3) receptors and is provided with analytical purity and stability data for proper storage and handling.
  • High affinity for 5-HT1B and 5-HT1D receptors (pKi 9.4 and 9.3).
  • Supplied as a powder with tested purity of 98.06%.
  • Available in small solid quantities and as a 10 mM solution in DMSO.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-42855