Learn More
TARGETMOL CHEMICALS INC DONITRIPTAN HYDROCHLORID 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2166625MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Donitriptan hydrochloride (Donitriptan monohydrochloride) is a potent high efficacy agonist of 5-HT1B/1D receptors with pKis of 9.4 and 9.3 respectivelypurity: 99%
Catalog No. 50-258-1014