Learn More
Immunoreagents Inc DONKEY A-GOAT IGG(H&L) BIO 2MG
Supplier:  Immunoreagents Inc DKXGT003DBIO
Encompass_Preferred
Secondary Antibody; Donkey anti-goat IgG (H&L), Affinity Pure, Biotin Conjugate, Host: Donkey, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-768