Learn More
Immunoreagents Inc DONKEY A-MOUSE IGG DYLIGHT594
Supplier:  Immunoreagents Inc DKXMU003F594NHSX
Encompass_Preferred
Secondary Antibody; Donkey anti-mouse IgG (H&L) - Affinity Pure, min x w/Bo, Ck, Eq, Gt, GP, Ha, Hu, Rb, Rt or Sh IgG, DyLight 594 Conjugate, Host: Donkey, Format: Lyo, Label: DyL594, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-928