Learn More
Immunoreagents Inc DONKEY A-RABBIT IGG HRP
Supplier:  Immunoreagents Inc DKXRB003FHRPX
Encompass_Preferred
Secondary Antibody; Donkey anti-rabbit IgG (H&L) - Affinity Pure, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Mu, Rt or Sh IgG, HRP Conjugate, Host: Donkey, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-063