Learn More
Immunoreagents Inc DONKEY A-SHEEP IGG DYLIGHT633
Supplier:  Immunoreagents Inc DKXSH003F633NHSX
Encompass_Preferred
Secondary Antibody: Donkey anti-sheep IgG (H&L) - Affinity Pure, min x w/Hu, Mu or Rb IgG, DyLight 633 Conjugate, Host: Donkey, Format: Lyo, Label: DyL633, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-964