Learn More
Cytek Biosciences Inc Donkey aMu IgG Anti Secondary
Supplier: Cytek Biosciences Inc 728090M001
Product Name: HRP Donkey Anti-Mouse IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: HRP Donkey Anti-Mouse IgG (H&L)
Cytek Catalog Number: 72-8090-M001
Marker: IgG (H&L)
Target Species: Mouse
Reported Cross Reactivity: Mouse
Clonality: Polyclonal
Host Species: Donkey
Conjugation: HRP
Size: 1 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.