Learn More
Jackson Immuno Research Labs Fluorescein (FITC) AffiniPure F(ab')₂ Fragment Donkey Anti-Chicken IgY (IgG) (H+L), .3mg
SDP
Supplier:  Jackson Immuno Research Labs 703096155
Encompass_Preferred
Fluorescein (FITC)-AffiniPure F(ab')2 Fragment Donkey Anti-Chicken IgY (IgG) (H+L) (min X Bov,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rb,Rat,Shp Sr Prot)
Catalog No. NC9663355