Learn More
Jackson Immuno Research Labs Alexa Fluor 647 AffiniPure Fab Fragment Donkey Anti-Mouse IgG (H+L), .75mg
SDP
Supplier:  Jackson Immuno Research Labs 715607003
Encompass_Preferred
Alexa Fluor 647-AffiniPure F(ab')2 Fragment Donkey Anti-Mouse IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Rb,Rat,Shp Sr Prot)
Catalog No. NC1204348