Learn More
MOLECULAR DEPOT LLC Donkey anti-Sheep IgG - 10 mg
Supplier:  MOLECULAR DEPOT LLC A2010045
Encompass_Preferred
Donkey anti-Sheep IgG (H L) is an affinity purified Donkey anti-Sheep IgG (H L) polyclonal antibody Highly pure and highly concentrated this antibody has several applications that include immunoassay development ELISA WB and immuno-histochemistry For in vitro research use only
Catalog No. 50-000-13404