Learn More
Alkali Scientific ASI™ Donor Goat Serum, 500mL
SDP
Supplier:  Alkali Scientific FB34
Encompass_Preferred
Donor Goat Serum is collected from controlled donor herds that receive regular veterinary inspection and care and are found free of hoofed animal diseases. Donor Goat Serum is a suitable alternative to fetal bovine serum in a wide range of cell culture applications such as the isolation and identification of viruses produced in a variety of primary cell lines. Virus-Screened Mycoplasma-Tested Triple 0.2 Micron Filtered This product ships on dry ice.
Catalog No. 50-302-9430