Learn More
Bio IVT Normal Human Serum, 1 mL, Frozen aliquot, IRB-approved, No clotting factors, Biomarker discovery, Diagnostic testing, Blood typing, Drug discovery, Cell biology
Supplier: Bio IVT SI13000011049
This product is a normal human serum provided by BIOIVT LLC, available in a 1 mL frozen aliquot. Ideal for a range of scientific applications, it serves as a valuable resource for biomarker discovery, diagnostic testing, and various cell biology and drug discovery studies. Collected under Institutional Review Board (IRB)-approved protocols, it ensures high quality and ethical sourcing, while being shipped overnight for immediate research needs.
- Source: Human
- Matrix: Serum
- State: Normal
- Volume: 1 mL
- Format: Frozen aliquot
- Collection protocol: IRB-approved
- Contains: Hormones, glucose, electrolytes, antibodies, antigens, nutrients
- Absence of: Clotting factors
- Applications: Biomarker discovery, diagnostic testing, blood typing, drug discovery, cell biology
- Availability: Readily available
- Shipping: Overnight shipping available
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.