Learn More
Cayman Chemical DonpezIl-d4hydrochlorIde 1mg
SDP
Supplier:  Cayman Chemical 182511 MG
Encompass_Preferred
An internal standard for the quantification of donepezil by GC- or LC-MS
Catalog No. 50-296-8065