Learn More
Medchemexpress LLC DOPA Decarboxylase Antibody (YA2428) | 50 UL

Supplier: Medchemexpress LLC HYP8268350UL
DOPA Decarboxylase Antibody (YA2428) is a Rabbit-derived, non-conjugated IgG monoclonal antibody that targets DOPA Decarboxylase, an enzyme crucial for neurotransmitter synthesis. DOPA Decarboxylase catalyzes the decarboxylation of L-3,4-dihydroxyphenylalanine (DOPA) to dopamine and L-5-hydroxytryptophan to serotonin. This antibody is intended for research use only.
- Rabbit-derived, non-conjugated IgG monoclonal antibody
- Targets DOPA decarboxylase
- Predicted and observed molecular weight of 54 kDa
- Reacts with human, mouse, and rat species
- Immunogen derived from human DOPA decarboxylase
- Suitable for western blot applications (1:500-1:1000 dilution)
- Endogenous sensitivity
- Purified by affinity chromatography
- Supplied as a liquid in a Tris-Glycine buffer with glycerol and BSA
- Stable for 1 year when stored at -20°C
- Shipped with blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.