Learn More
Cayman Chemical DopamIn-d4hydrochlorIde 25mg
SDP
Supplier:  Cayman Chemical 3630925 MG
Encompass_Preferred
An internal standard for the quantification of dopamine by GC- or LC-MS
Catalog No. 50-291-3067