Learn More
Cayman Chemical DopamIn-d4hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 363095 MG
Encompass_Preferred
An internal standard for the quantification of dopamine by GC- or LC-MS
Catalog No. 50-291-2946