Learn More
Medchemexpress LLC Dopamine-13c,15n (hydrochloride) | 369656-74-6 | MFCD01074386 | 99.0% | 191.63 g/mol | C7[13C]H12[15N]O2 · HCl | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0451AS610MG
Encompass_Preferred
Dopamine-13C,15N hydrochloride is the 13C- and 15N-labeled isotopologue of dopamine hydrochloride used as a stable-isotope standard and tracer in analytical, metabolic, and quantitative mass spectrometry studies. It is supplied as a solid hydrochloride salt with high analytical purity and is intended for laboratory use in trace-level analyses.
  • Isotopically labeled internal standard for mass spectrometry
  • High analytical purity (≈99.0%)
  • Supplied as a solid hydrochloride salt for stability
  • Available in small milligram quantities for analytical workflows
  • Store sealed at -20°C and protect from moisture
Catalog No. 50-002-99023