Learn More
Medchemexpress LLC Dopamine-d3 hydrochloride | 53587-30-7 | MFCD03428087 | 98.0% | 192.66 g/mol | C8H9D3ClNO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0451AS410MG
Encompass_Preferred
Dopamine-d3 hydrochloride is a deuterium-labeled analogue of dopamine supplied as the hydrochloride salt for use as an internal standard or tracer in analytical and research applications, including mass spectrometry and isotope-labeling studies.
  • Deuterium-labeled (d3) form for accurate mass spectrometry quantification.
  • Hydrochloride salt, stable solid suitable for analytical workflows.
  • CAS number 53587-30-7 and molecular weight 192.66 g/mol.
  • High isotopic enrichment and chemical purity for reliable internal standard performance.
  • Suitable for preparation of calibration standards and method validation in bioanalysis.
Catalog No. 50-002-99024