Learn More
Biomatik Corporation Dopamine (DA) CLIA Kit
Supplier:  Biomatik Corporation EKU08644
Encompass_Preferred
Dopamine (DA) CLIA Kit for detecting DA in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 3.9-1,000pg/ml. Sensitivity: 1.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: 2-(3,4-Dihydroxyphenyl) ethylamine; 3,4-Dihydroxyphenethylamine; 3-Hydroxytyramine; Intropin; Revivan; Oxytyramine; Dopamine Hydrochloride. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4418