Learn More
Medchemexpress LLC Dopamine-PEG-N3 MW 100mg
SDP
Supplier:  Medchemexpress LLC HYW1123941C100MG
Encompass_Preferred
Dopamine-PEG-N3 (MW 2000) (Dopamine-PEG-Azide (MW 2000)) is a compound composed of dopamine PEG segments and an azide group (-N3) Dopamine-PEG-N3 (MW 2000) can be used as a crosslinker to connect different materials together to build new composite materials and improve the performance of materials Dopamine-PEG-N3 (MW 2000) is a click chemistry reagent It contains an azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing an alkyne group
Catalog No. 50-002-06101