Learn More
Medchemexpress LLC DOPE-GA | 228706-30-7 | C46H84NO11P | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15024050MG
Encompass_Preferred
DOPE-GA is used in the preparation of liposomes for drug delivery research and other applications. It is a key raw material for preparing liposome ELL-12, a non-myelosuppressive antiproliferative agent with anti-tumor activity.
  • Affects the structure and stability of liposomes
  • Influences the interaction between ELL-12 and cells
  • Oil appearance, colorless to light yellow
  • Purity: ≥98.0%
Catalog No. 50-003-75327