Learn More
Medchemexpress LLC Dope-mal | 2295813-15-7 | 98.7% | 895.15 | C48H83N2O11P | 10mg
SDP
Supplier:  Medchemexpress LLC HYW80080510MG
Encompass_Preferred
DOPE-Mal is a synthetic analog of naturally-occurring PE containing 18 1 fatty acids at the sn-1 and sn-2 positions with a terminal maliemide group The maleimide group will react with a thiol group to form a covalent bond The hydrophilic PEG spacer increases solubility in aqueous media
Catalog No. 50-002-68358