Learn More
Medchemexpress LLC DOPE-NHS | 1010188-79-0 | C50H87N2O13P | 100 MG
SDP
Supplier:  Medchemexpress LLC HY150241100MG
Encompass_Preferred
DOPE-NHS is a linker used for conjugating peptides to exosomes and other membrane-based nanoparticles. It also finds applications in drug delivery, enabling various research applications.
  • Used for conjugating peptides to exosomes and other membrane-based nanoparticles.
  • Applicable for drug delivery purposes.
  • Facilitates conjugation of peptides to exosomes and other membrane-based nanoparticles in vitro.
  • Can be used to conjugate MSC derived exosomes with rabies viral glycoprotein.
  • Can be used to conjugate exosomes with cardiac homing peptide.
Catalog No. 50-003-75581