Learn More
Medchemexpress LLC Dope-peg-azide | 99.8% | 3,400 Da (average) | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW591450500MG
Encompass_Preferred
DOPE-PEG-Azide (MW 3400) is a PEGylated phospholipid in which dioleoylphosphatidylethanolamine (DOPE) is linked to a polyethylene glycol chain terminating in an azide group. Supplied as a powder (average MW 3,400 Da), it provides an azide functional handle for bioorthogonal conjugation and for surface modification of liposomes and nanoparticles.
  • Azide-terminated PEG chain for bioorthogonal click chemistry.
  • Average molecular weight 3,400 Da (polydisperse PEG).
  • Supplied as a powder with recommended cold storage for stability.
  • Suitable for liposome and nanoparticle surface functionalization.
  • High purity (99.8%) appropriate for research use.
Catalog No. 50-002-84776