Learn More
Medchemexpress LLC Dope-peg-dbco | 90.0% | 2000 (average) | 10 MG

Supplier: Medchemexpress LLC HY16403510MG
DOPE-PEG-DBCO (MW 2000) is a functionalized PEG-lipid in which DOPE is linked through a polyethylene glycol spacer (average MW 2000) to a dibenzocyclooctyne (DBCO) moiety. It is used to introduce a bioorthogonal DBCO handle into lipid assemblies, enabling strain-promoted azide-alkyne cycloaddition (SPAAC) for bioconjugation and surface functionalization.
- Average molecular weight ~2000 Da.
- Purity approximately 90.0%.
- Provides a DBCO handle for SPAAC click chemistry.
- DOPE supports membrane fusion and acts as a helper lipid.
- PEG spacer offers steric stabilization of lipid particles.
- Soluble in DMSO at ~100 mg/mL (requires ultrasonic).
- Recommended storage: -20°C; in solvent: -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.