Learn More
Medchemexpress LLC Dopexamine hydrochloride | 86484-91-5 | 429.42 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYU0020510MM/1ML
Encompass_Preferred
Dopexamine hydrochloride is a β2 adrenergic receptor agonist and a dopamine analogue. It exhibits agonist activity at both β2 and dopaminergic receptors. This compound can attenuate systemic inflammatory responses, reduce tissue leukocyte infiltration, and protect against organ injury, even at doses that do not impact global hemodynamics or regional microvascular flow.
  • Attenuates systemic inflammatory response.
  • Reduces tissue leukocyte infiltration.
  • Protects against organ injury.
  • Maintains global hemodynamics and regional microvascular flow.
  • Reduces systemic inflammatory response to endotoxin.
  • Impacts cytokine release, endothelial adhesion molecules, and oxidative stress.
Catalog No. 50-004-46911