Learn More
Medchemexpress LLC Dopexamine hydrochloride | 86484-91-5 | 429.42 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYU00205100MG
Encompass_Preferred
Dopexamine hydrochloride is a β2 adrenergic receptor agonist, intended for research use only. This dopamine analogue exhibits agonist activity at β2 and dopaminergic receptors, making it suitable for studies on inflammatory response and organ protection.
  • Attenuates systemic inflammatory response
  • Reduces tissue leukocyte infiltration
  • Protects against organ injury
  • Reduces systemic inflammatory response to endotoxin, including cytokine release, endothelial adhesion molecules, and oxidative stress
Catalog No. 50-004-46941