Learn More
Medchemexpress LLC Dopexamine hydrochloride | 86484-91-5 | 429.42 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYU0020550MG
Encompass_Preferred
Dopexamine hydrochloride is a β2 adrenergic receptor agonist and dopamine analogue. It acts on β2 and dopaminergic receptors to attenuate systemic inflammatory response, reduce tissue leukocyte infiltration, and protect against organ injury. This compound helps reduce inflammation, cytokine release, endothelial adhesion molecules, and oxidative stress without significantly altering blood pressure or stroke volume.
  • Attenuates systemic inflammatory response
  • Reduces tissue leukocyte infiltration
  • Protects against organ injury
  • Reduces systemic inflammatory response to endotoxin
  • Reduces cytokine release
  • Reduces endothelial adhesion molecules
  • Reduces oxidative stress
  • Does not substantially change systemic hemodynamics
Catalog No. 50-004-49663